Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fndc5
Entrez Gene ID
100757300
Gene Name
fibronectin type III domain containing 5
Gene Aliases
I79_006895
Chromosome Location
Chr.Un|NW_003613801: 1317444 - 1335613 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500650.1 XP_003500698.1 1 - 2 104 59

View Full Product Details