Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tceal7
Entrez Gene ID
100771506
Gene Name
transcription elongation factor A (SII)-like 7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614444: 106220 - 106516 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507899.1 XP_003507947.1 1 - 1 215 128

View Full Product Details