Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zdhhc24
Entrez Gene ID
100760813
Gene Name
zinc finger, DHHC-type containing 24
Gene Aliases
I79_018382
Chromosome Location
Chr.Un|NW_003614834: 6234 - 13948 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510148.2 XP_003510196.1 2 - 3 580 98

View Full Product Details