Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcnk9
Entrez Gene ID
100756282
Gene Name
potassium channel, subfamily K, member 9
Gene Aliases
I79_019162
Chromosome Location
Chr.Un|NW_003614962: 181292 - 220157 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510774.1 XP_003510822.1 1 - 2 284 71

View Full Product Details