Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Trim65
Entrez Gene ID
100758291
Gene Name
tripartite motif containing 65
Gene Aliases
I79_014335
Chromosome Location
Chr.Un|NW_003614300: 210726 - 216029 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647376.1 XP_007645566.1 4 - 5 396 103

View Full Product Details