Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vsnl1
Entrez Gene ID
100757699
Gene Name
visinin-like 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613936: 565610 - 614949 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643664.1 XP_007641854.1 3 - 4 503 81

View Full Product Details