Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lrrc24
Entrez Gene ID
100759630
Gene Name
leucine rich repeat containing 24
Gene Aliases
I79_015336
Chromosome Location
Chr.Un|NW_003614411: 68504 - 71951 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507650.2 XP_003507698.1 1 - 2 257 106

View Full Product Details