Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ntn3
Entrez Gene ID
100771322
Gene Name
netrin 3
Gene Aliases
Netrin-3
Chromosome Location
Chr.Un|NW_003614722: 284147 - 286703 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509532.2 XP_003509580.1 5 - 6 1540 59

View Full Product Details