Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam71b
Entrez Gene ID
100767497
Gene Name
family with sequence similarity 71, member B
Gene Aliases
I79_018342
Chromosome Location
Chr.Un|NW_003614829: 243314 - 246261 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510089.1 XP_003510137.1 2 - 3 613 97

View Full Product Details