Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bola2b
Entrez Gene ID
100770374
Gene Name
bolA family member 2B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614830: 1963 - 3220 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510098.2 XP_003510146.1 2 - 3 809 71

View Full Product Details