Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc22a11
Entrez Gene ID
100769171
Gene Name
solute carrier family 22 (organic anion/urate transporter), member 11
Gene Aliases
I79_025007
Chromosome Location
Chr.Un|NW_003618118: 8895 - 23148 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515456.1 XP_003515504.1 4 - 5 817 57

View Full Product Details