Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ndnf
Entrez Gene ID
100757334
Gene Name
neuron-derived neurotrophic factor
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614267: 647645 - 657908 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506633.1 XP_003506681.1 2 - 3 317 94

View Full Product Details