Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Alx3
Entrez Gene ID
100770020
Gene Name
ALX homeobox 3
Gene Aliases
I79_022707
Chromosome Location
Chr.Un|NW_003616075: 105800 - 114723 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513692.2 XP_003513740.1 1 - 2 31 66

View Full Product Details