Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfap2e
Entrez Gene ID
100761645
Gene Name
transcription factor AP-2 epsilon (activating enhancer binding protein 2 epsilon)
Gene Aliases
I79_007180
Chromosome Location
Chr.Un|NW_003613817: 1067055 - 1083596 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641625.1 XP_007639815.1 5 - 6 689 69

View Full Product Details