Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hes3
Entrez Gene ID
100754865
Gene Name
hes family bHLH transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616989: 351 - 1539 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514806.2 XP_003514854.2 1 - 2 62 66

View Full Product Details