Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ca1
Entrez Gene ID
100772227
Gene Name
carbonic anhydrase I
Gene Aliases
I79_004473
Chromosome Location
Chr.Un|NW_003613698: 2210569 - 2219209 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498597.1 XP_003498645.1 2 - 3 240 121

View Full Product Details