Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wipf3
Entrez Gene ID
100759347
Gene Name
WAS/WASL interacting protein family, member 3
Gene Aliases
I79_015522
Chromosome Location
Chr.Un|NW_003614428: 256543 - 307470 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507793.1 XP_003507841.1 5 - 6 678 71

View Full Product Details