Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map7d2
Entrez Gene ID
100752405
Gene Name
MAP7 domain containing 2
Gene Aliases
I79_005596, Mtap7d2
Chromosome Location
Chr.Un|NW_003613740: 1621019 - 1674393 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640253.1 XP_007638443.1 12 - 13 1630 103

View Full Product Details