Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfam1
Entrez Gene ID
100752842
Gene Name
NFAT activating protein with ITAM motif 1
Gene Aliases
I79_016826
Chromosome Location
Chr.Un|NW_003614589: 205647 - 236089 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649419.1 XP_007647609.1 3 - 4 482 70

View Full Product Details