Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc35b2
Entrez Gene ID
100758269
Gene Name
solute carrier family 35 (adenosine 3'-phospho 5'-phosphosulfate transporter), member B2
Gene Aliases
I79_008087
Chromosome Location
Chr.Un|NW_003613855: 1359906 - 1363282 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642329.1 XP_007640519.1 1 - 2 316 92

View Full Product Details