Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myf5
Entrez Gene ID
100773160
Gene Name
myogenic factor 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614543: 523168 - 525774 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508435.2 XP_003508483.1 1 - 2 664 80

View Full Product Details