Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam83b
Entrez Gene ID
100761983
Gene Name
family with sequence similarity 83, member B
Gene Aliases
I79_000112
Chromosome Location
Chr.Un|NW_003613582: 103097 - 152737 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495007.1 XP_003495055.1 2 - 3 611 106

View Full Product Details