Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tmem52
Entrez Gene ID
100755990
Gene Name
transmembrane protein 52
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614679: 124609 - 125955 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509281.1 XP_003509329.1 2 - 3 117 93

View Full Product Details