Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tarm1
Entrez Gene ID
100764097
Gene Name
T cell-interacting, activating receptor on myeloid cells 1
Gene Aliases
I79_017844
Chromosome Location
Chr.Un|NW_003614747: 314931 - 328829 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650269.1 XP_007648459.1 2 - 3 333 71

View Full Product Details