Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cd33
Entrez Gene ID
100753674
Gene Name
CD33 molecule
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615616: 80162 - 81989 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007653248.1 XP_007651438.1 1 - 2 253 66

View Full Product Details