Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdr9c7
Entrez Gene ID
100761860
Gene Name
short chain dehydrogenase/reductase family 9C, member 7
Gene Aliases
I79_013469
Chromosome Location
Chr.Un|NW_003614217: 40553 - 52441 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506172.1 XP_003506220.1 2 - 3 561 63

View Full Product Details