Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdcp2
Entrez Gene ID
100770767
Gene Name
CUB domain containing protein 2
Gene Aliases
I79_001004
Chromosome Location
Chr.Un|NW_003613599: 3766739 - 3783884 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643825.1 XP_007642015.1 4 - 5 1121 93

View Full Product Details