Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpp14
Entrez Gene ID
100769100
Gene Name
ribonuclease P/MRP 14kDa subunit
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613911: 631117 - 638070 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502729.2 XP_003502777.2 2 - 3 308 83
XM_007643345.1 XP_007641535.1 2 - 3 211 83
XM_007643346.1 XP_007641536.1 2 - 3 111 83
GenBank mRNA JP054914.1 - 1 - 2 103 83

View Full Product Details