Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bik
Entrez Gene ID
100768268
Gene Name
BCL2-interacting killer (apoptosis-inducing)
Gene Aliases
I79_015850
Chromosome Location
Chr.Un|NW_003614471: 333014 - 337451 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508026.2 XP_003508074.1 1 - 2 657 87

View Full Product Details