Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Synpo2l
Entrez Gene ID
100757374
Gene Name
synaptopodin 2-like
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613632: 76105 - 83709 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496719.1 XP_003496767.1 1 - 2 108 76

View Full Product Details