Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rundc1
Entrez Gene ID
100752621
Gene Name
RUN domain containing 1
Gene Aliases
I79_011743
Chromosome Location
Chr.Un|NW_003614078: 1087777 - 1098693 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504635.2 XP_003504683.1 5 - 6 919 72
GenBank mRNA JI862594.1 - 3 - 4 398 72
JP045997.1 - 3 - 4 398 72

View Full Product Details