Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Atp6v1g2
Entrez Gene ID
100765359
Gene Name
ATPase, H+ transporting, lysosomal 13kDa, V1 subunit G2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614548: 617816 - 620919 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508489.2 XP_003508537.1 2 - 3 345 93

View Full Product Details