Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Amtn
Entrez Gene ID
100768466
Gene Name
amelotin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615639: 44396 - 55698 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512878.1 XP_003512926.1 6 - 7 353 72

View Full Product Details