Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC100758463
Entrez Gene ID
100758463
Gene Name
chromosome unknown open reading frame, human C15orf60
Gene Aliases
I79_008976
Chromosome Location
Chr.Un|NW_003613894: 933603 - 1016775 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502409.2 XP_003502457.1 3 - 4 331 76

View Full Product Details