Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Id3
Entrez Gene ID
100751577
Gene Name
inhibitor of DNA binding 3, dominant negative helix-loop-helix protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614105: 149914 - 151477 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504920.2 XP_003504968.1 1 - 2 370 74

View Full Product Details