Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEA15
Entrez Gene ID
102185324
Gene Name
proliferation and apoptosis adaptor protein 15
Gene Aliases
-
Chromosome Location
Chr.3: 110454014 - 110464196 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046375.1 XP_017901864.1 2 - 3 337 74
GenBank mRNA JO355652.2 - 2 - 3 280 74
JO557227.2 - 2 - 3 316 74
JO633971.2 - 2 - 3 298 74

View Full Product Details