Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UBE2D2
Entrez Gene ID
102171866
Gene Name
ubiquitin conjugating enzyme E2 D2
Gene Aliases
-
Chromosome Location
Chr.7: 59671313 - 59714770 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018050305.1 XP_017905794.1 6 - 7 1026 92

View Full Product Details