Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAV2
Entrez Gene ID
102176581
Gene Name
caveolin 2
Gene Aliases
-
Chromosome Location
Chr.4: 68384847 - 68392320 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013963312.2 XP_013818766.1 2 - 3 486 93
XM_018047184.1 XP_017902673.1 2 - 3 319 93
GenBank mRNA JO394152.2 - 2 - 3 552 93

View Full Product Details