Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C21H15orf40
Entrez Gene ID
102178861
Gene Name
chromosome 21 C15orf40 homolog
Gene Aliases
-
Chromosome Location
Chr.21: 22550223 - 22555795 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695017.3 XP_005695074.1 3 - 4 612 99

View Full Product Details