Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKAB1
Entrez Gene ID
102181705
Gene Name
protein kinase AMP-activated non-catalytic subunit beta 1
Gene Aliases
-
Chromosome Location
Chr.17: 15483196 - 15493757 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013970630.2 XP_013826084.1 6 - 7 872 92
XM_013970631.2 XP_013826085.1 6 - 7 944 92
XM_013970632.2 XP_013826086.1 6 - 7 810 92

View Full Product Details