Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LHB
Entrez Gene ID
102180531
Gene Name
luteinizing hormone beta polypeptide
Gene Aliases
LHbeta
Chromosome Location
Chr.18: 56589042 - 56590579 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062755.1 XP_017918244.1 2 - 3 671 56
GenBank mRNA KA349409.1 - 1 - 2 472 56
KM054525.1 - 2 - 3 185 56
KP310924.1 - 2 - 3 185 56
KP310925.1 - 2 - 3 185 56

View Full Product Details