Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CKMT1A
Entrez Gene ID
102183273
Gene Name
creatine kinase, mitochondrial 1A
Gene Aliases
CKMT1B
Chromosome Location
Chr.21: 54833207 - 54839469 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695305.3 XP_005695362.1 3 - 4 625 70
XM_018066415.1 XP_017921904.1 3 - 4 931 70
XM_018066418.1 XP_017921907.1 3 - 4 615 70
XM_018066419.1 XP_017921908.1 3 - 4 632 70
GenBank mRNA KA309735.1 - 3 - 4 718 70

View Full Product Details