Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GABARAP
Entrez Gene ID
102182439
Gene Name
GABA type A receptor-associated protein
Gene Aliases
-
Chromosome Location
Chr.19: 26765219 - 26767279 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693485.3 XP_005693542.1 3 - 4 444 94
GenBank mRNA JO581681.1 - 3 - 4 1006 94

View Full Product Details