Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GADD45A
Entrez Gene ID
102179904
Gene Name
growth arrest and DNA damage inducible alpha
Gene Aliases
-
Chromosome Location
Chr.3: 43245994 - 43249270 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678467.3 XP_005678524.2 2 - 3 517 80
GenBank mRNA JO376216.2 - 2 - 3 754 80

View Full Product Details