Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCNP
Entrez Gene ID
102169907
Gene Name
PEST proteolytic signal containing nuclear protein
Gene Aliases
-
Chromosome Location
Chr.1: 45740750 - 45755856 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013975550.2 XP_013831004.2 3 - 4 750 74
GenBank mRNA JO324224.2 - 2 - 3 309 74
JO324225.2 - 2 - 3 309 74
JO363573.2 - 3 - 4 405 74
JO394550.2 - 2 - 3 481 74
JO451355.2 - 2 - 3 301 74
JO498946.2 - - 616 74
JO508115.2 - 1 - 2 222 74
JO556981.2 - 3 - 4 435 74
JO556982.2 - 3 - 4 432 74
JO610616.2 - 2 - 3 300 74
KA310411.1 - 2 - 3 1004 74

View Full Product Details