Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTF2H5
Entrez Gene ID
102180044
Gene Name
general transcription factor IIH subunit 5
Gene Aliases
-
Chromosome Location
Chr.9: 82017605 - 82030018 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684941.3 XP_005684998.1 2 - 3 154 85
XM_013966630.2 XP_013822084.1 2 - 3 94 85
GenBank mRNA KA342928.1 - 1 - 2 382 85

View Full Product Details