Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COMMD1
Entrez Gene ID
102172634
Gene Name
copper metabolism domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 60179779 - 60348796 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686785.3 XP_005686842.1 1 - 2 213 71

View Full Product Details