Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LYPD4
Entrez Gene ID
102176159
Gene Name
LY6/PLAUR domain containing 4
Gene Aliases
-
Chromosome Location
Chr.18: 52273399 - 52282688 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692577.2 XP_005692634.1 3 - 4 372 82

View Full Product Details