Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
KLHL21
Entrez Gene ID
102180066
Gene Name
kelch like family member 21
Gene Aliases
-
Chromosome Location
Chr.16: 45337816 - 45350888 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018060419.1 XP_017915908.1 1 - 2 1124 68

View Full Product Details