Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNG2
Entrez Gene ID
102174727
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 2
Gene Aliases
-
Chromosome Location
Chr.5: 73670713 - 73788601 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680662.3 XP_005680719.1 1 - 2 1558 79
GenBank mRNA JO635545.2 - 1 - 2 128 79

View Full Product Details